339385

Sigma-Aldrich

4,4-Diphenyl-2-cyclohexen-1-one

99%

Linear Formula:
(C6H5)2C6H6(=O)
CAS Number:
Molecular Weight:
248.32
MDL number:
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

assay

99%

mp

90-92 °C (lit.)

SMILES string

O=C1CCC(C=C1)(c2ccccc2)c3ccccc3

InChI

1S/C18H16O/c19-17-11-13-18(14-12-17,15-7-3-1-4-8-15)16-9-5-2-6-10-16/h1-11,13H,12,14H2

InChI key

LUFGFXHHSGISSL-UHFFFAOYSA-N

Related Categories

General description

Photochemistry of 4,4-diphenyl-2-cyclohepten-1-one has been investigated. Electrohydrodimerization of 4,4-diphenyl-2-cyclohexen-1-one in buffered hydroethanolic solutions has been studied by polarography, cyclic voltammetry and controlled-potential coulometry.

Packaging

1 g in glass bottle

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Electrochemical reduction of 2-cyclohexen-1-ones in a hydroethanolic medium.
Brillas E and Ortiz A.
J. Chem. Soc., Faraday, 82(2), 495-508 (1986)
Photochemistry of cis-4, 4-diphenyl-2-cyclohepten-1-one.
Bunce RA, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 57(1), 317-329 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.