339547

Sigma-Aldrich

3-Bromo-4-fluorobenzaldehyde

98%

Linear Formula:
BrC6H3(F)CHO
CAS Number:
Molecular Weight:
203.01
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

assay

98%

bp

138-139 °C/2.5 mmHg (lit.)

mp

31-33 °C (lit.)

SMILES string

Fc1ccc(C=O)cc1Br

InChI

1S/C7H4BrFO/c8-6-3-5(4-10)1-2-7(6)9/h1-4H

InChI key

FAHZIKXYYRGSHF-UHFFFAOYSA-N

Application

3-Bromo-4-fluorobenzaldehyde was used in the synthesis of 4-fluoro-3-phenoxybenzoic acid-glycine. It was also used in the synthesis of Schiff bases of 4-nitroaniline/ 2,4-dinitroaniline/3,4-dichloroaniline/4-methoxy-3-nitro aniline via ultrasound irradiation method.

Packaging

5 g in glass bottle

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xn

Risk Statement

22-36/37/38

Safety Statement

26-36

RIDADR

NONH for all modes of transport

WGK Germany

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Ultrasonic assisted synthesis, acoustical property and antibacterial activity of some Schiff bases.
Premalatha R and Santhi N.
International Letters of Chemistry, Physics and Astronomy, 14(1), 53-64 (2014)
Guomin Shan et al.
Chemical research in toxicology, 17(2), 218-225 (2004-02-18)
The pyrethroids are one of the most heavily used insecticide classes in the world. It is important to develop sensitive and rapid analytical techniques for environmental monitoring and assessment of human exposure to these compounds. Because major pyrethroids contain a...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.