All Photos(1)

340510

Sigma-Aldrich

sec-Butyl tiglate

97%

Linear Formula:
CH3CH=C(CH3)CO2CH(CH3)C2H5
CAS Number:
Molecular Weight:
156.22
MDL number:
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.435 (lit.)

bp

84.5 °C/27 mmHg (lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

CCC(C)OC(=O)\C(C)=C\C

InChI

1S/C9H16O2/c1-5-7(3)9(10)11-8(4)6-2/h5,8H,6H2,1-4H3/b7-5+

InChI key

UKJTZSWULQNVJI-FNORWQNLSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service