341819

Sigma-Aldrich

Cobalt(II) thiocyanate hydrate

Empirical Formula (Hill Notation):
C2CoN2S2 · xH2O
CAS Number:
Molecular Weight:
175.10 (anhydrous basis)
EC Number:
MDL number:

SMILES string

O.N#CS[Co]SC#N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service