Documents

342173

Sigma-Aldrich

Tin(II) iodide

95%

Synonym(s):
Tin diiodide, Stannous iodide
Linear Formula:
SnI2
CAS Number:
Molecular Weight:
372.52
EC Number:
MDL number:

assay

95%

bp

714 °C (lit.)

mp

320 °C (lit.)

density

5.28 g/mL at 25 °C (lit.)

SMILES string

I[SnH2]I

InChI

1S/2HI.Sn/h2*1H;/q;;+2/p-2

InChI key

JTDNNCYXCFHBGG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3260 8 / PGII

WGK Germany

3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.