Documents

344117

Sigma-Aldrich

(S)-(−)-N,N-Dimethyl-1-ferrocenylethylamine

≥95%

Synonym(s):
(S)-(−)-[1-(Dimethylamino)ethyl]ferrocene
Empirical Formula (Hill Notation):
C14H19FeN
CAS Number:
Molecular Weight:
257.15
MDL number:
PubChem Substance ID:

assay

≥95%

optical activity

[α]20/D −13°, c = 1 in ethanol (5 h)

bp

120-121 °C/0.7 mmHg (lit.)

density

1.222 g/mL at 20 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][CH]2)N(C)C

InChI

1S/C9H14N.C5H5.Fe/c1-8(10(2)3)9-6-4-5-7-9;1-2-4-5-3-1;/h4-8H,1-3H3;1-5H;/t8-;;/m0../s1

InChI key

ISOLNZQTVMCEOI-JZGIKJSDSA-N

Looking for similar products? Visit Product Comparison Guide

RIDADR

NONH for all modes of transport

WGK Germany

3

Flash Point(F)

203 °F

Flash Point(C)

95 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.