MilliporeSigma
All Photos(1)

344605

Sigma-Aldrich

4-Fluoro-3-methylphenylmagnesium bromide solution

1.0 M in THF

Linear Formula:
FC6H3(CH3)MgBr
CAS Number:
Molecular Weight:
213.33
MDL number:
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

density

1.001 g/mL at 25 °C

SMILES string

Cc1cc([Mg]Br)ccc1F

InChI

1S/C7H6F.BrH.Mg/c1-6-4-2-3-5-7(6)8;;/h3-5H,1H3;1H;/q;;+1/p-1

InChI key

OLSGDQISPRACFY-UHFFFAOYSA-M

Related Categories

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service