Documents

344850

Sigma-Aldrich

4,4′-Trimethylenedipiperidine dithiocyanate

98%

Empirical Formula (Hill Notation):
C14H27N3S
CAS Number:
Molecular Weight:
269.45
EC Number:
MDL number:

assay

98%

mp

209-211 °C (lit.)

SMILES string

SC#N.SC#N.C(CC1CCNCC1)CC2CCNCC2

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service