345024

Sigma-Aldrich

2-(p-Toluoyl)benzoic acid

98%

Linear Formula:
CH3C6H4COC6H4CO2H
CAS Number:
Molecular Weight:
240.25
EC Number:
MDL number:

assay

98%

mp

137-139 °C (lit.)

SMILES string

Cc1ccc(cc1)C(=O)c2ccccc2C(O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service