347841

Sigma-Aldrich

1,9-Decadiene

96%

Linear Formula:
CH2=CH(CH2)6CH=CH2
CAS Number:
Molecular Weight:
138.25
Beilstein/REAXYS Number:
1697870
EC Number:
MDL number:
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.432 (lit.)

bp

169 °C (lit.)

density

0.75 g/mL at 25 °C (lit.)

SMILES string

C=CCCCCCCC=C

InChI

1S/C10H18/c1-3-5-7-9-10-8-6-4-2/h3-4H,1-2,5-10H2

InChI key

NLDGJRWPPOSWLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Precautionary Statements

hazcat

Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

storage_class_code

3 - Flammable liquids

WGK Germany

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service