348910

Sigma-Aldrich

(−)-Dimenthyl fumarate

95%

Synonym(s):
(−)-Di[(1R)-menthyl] fumarate
Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Beilstein/REAXYS Number:
2568525
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

95%

optical activity

[α]20/D −102°, c = 1 in chloroform

bp

211 °C/5 mmHg (lit.)

mp

59-61 °C (lit.)

SMILES string

C[C@@H]1CC[C@@H](C(C)C)[C@@H](C1)OC(=O)\C=C\C(=O)O[C@@H]2C[C@H](C)CC[C@H]2C(C)C

InChI

1S/C24H40O4/c1-15(2)19-9-7-17(5)13-21(19)27-23(25)11-12-24(26)28-22-14-18(6)8-10-20(22)16(3)4/h11-12,15-22H,7-10,13-14H2,1-6H3/b12-11+/t17-,18-,19+,20+,21-,22-/m1/s1

InChI key

HTJZKHLYRXPLLS-NGHDTEGZSA-N

Related Categories

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.