Documents

357812

Sigma-Aldrich

4-(1-Hydroxy-1-methylethyl)-2,2,5,5-tetramethyl-3-imidazolinium-1-yloxy, free radical

98%

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
MDL number:
PubChem Substance ID:

assay

98%

mp

155-157 °C (lit.)

SMILES string

CC(C)(O)C1=NC(C)(C)N([O])C1(C)C

InChI

1S/C10H19N2O2/c1-8(2)7(9(3,4)13)11-10(5,6)12(8)14/h13H,1-6H3

InChI key

YXJJGNAHYNJTNP-UHFFFAOYSA-N

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.