Documents

365211

Sigma-Aldrich

Lithium iodoacetate

97%

Synonym(s):
Iodoacetic acid lithium salt
Linear Formula:
ICH2COOLi
CAS Number:
Molecular Weight:
191.88
Beilstein/REAXYS Number:
4208524
EC Number:
MDL number:

assay

97%

mp

239 °C (dec.) (lit.)

SMILES string

[Li+].[O-]C(=O)CI

InChI

1S/C2H3IO2.Li/c3-1-2(4)5;/h1H2,(H,4,5);/q;+1/p-1

InChI key

RZCFQIDTJPHHFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service