Skip to Content
MilliporeSigma

Skip To

365688

N,N--Dimethyloctylamine N-oxide

99%

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CH3(CH2)7N(O)(CH3)2
CAS Number:
Molecular Weight:
173.30
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
2039368


assay

99%

mp

128-130 °C (lit.)

SMILES string

CCCCCCCC[N+](C)(C)[O-]



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library