Documents

366781

Sigma-Aldrich

4,4′-Diheptylazoxybenzene

liquid crystal (nematic)

Linear Formula:
CH3(CH2)6C6H4N=N(O)C6H4(CH2)6CH3
CAS Number:
Molecular Weight:
394.59
MDL number:
PubChem Substance ID:

form

liquid crystal (nematic)

SMILES string

CCCCCCCc1ccc(cc1)\N=[N+](/[O-])c2ccc(CCCCCCC)cc2

WGK Germany

3

Flash Point(F)

235 °F

Flash Point(C)

113 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.