All Photos(1)

367311

Sigma-Aldrich

2-(2-Bromoethyl)-2,5,5-trimethyl-1,3-dioxane

95%

Empirical Formula (Hill Notation):
C9H17BrO2
CAS Number:
Molecular Weight:
237.13
Beilstein:
4305359
MDL number:

assay

95%

refractive index

n20/D 1.470 (lit.)

bp

72-73 °C/0.6 mmHg (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

CC1(C)COC(C)(CCBr)OC1

InChI

1S/C9H17BrO2/c1-8(2)6-11-9(3,4-5-10)12-7-8/h4-7H2,1-3H3

InChI key

HSWLDQSQVWEZIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service