Documents

367680

1,1,1,3,3,3-Hexafluoro-2,2-bis(4-methylphenyl)propane

99%

Synonym(s):
2,2-Di(p-tolyl)-1,1,1,3,3,3-trifluoropropane, Hexafluoro-2,2-bis(4-methylphenyl)propane
Empirical Formula (Hill Notation):
C17H14F6
CAS Number:
Molecular Weight:
332.28
MDL number:

assay

99%

SMILES string

Cc1ccc(cc1)C(c2ccc(C)cc2)(C(F)(F)F)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service