All Photos(1)

367966

3-Chloro-2,4-difluoroaniline

99%

Empirical Formula (Hill Notation):
C6H4ClF2N
CAS Number:
Molecular Weight:
163.55
MDL number:

assay

99%

SMILES string

Nc1ccc(F)c(Cl)c1F

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service