Skip to Content
MilliporeSigma

371386

4,4′-Dihydroxybiphenyl

purified by sublimation, ≥99%

Synonym(s):

4,4′-Biphenol, 4,4′-Biphenyldiol, 4,4′-Diphenol

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
HOC6H4C6H4OH
CAS Number:
Molecular Weight:
186.21
EC Number:
202-200-5
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1908886
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

VCCBEIPGXKNHFW-UHFFFAOYSA-N

InChI

1S/C12H10O2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,13-14H

SMILES string

Oc1ccc(cc1)-c2ccc(O)cc2

assay

≥99%

purified by

sublimation

mp

280-282 °C (lit.)

Gene Information

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service