373982

Sigma-Aldrich

Bis(5-chloro-2-hydroxyphenyl)methane

technical grade, 90%

Synonym(s):
Dichlorophene, Dichlorophen
Linear Formula:
CH2[C6H3(Cl)OH]2
CAS Number:
Molecular Weight:
269.12
Beilstein/REAXYS Number:
1884514
EC Number:
MDL number:
PubChem Substance ID:

grade

technical grade

assay

90%

mp

168-172 °C (lit.)

SMILES string

Oc1ccc(Cl)cc1Cc2cc(Cl)ccc2O

InChI

1S/C13H10Cl2O2/c14-10-1-3-12(16)8(6-10)5-9-7-11(15)2-4-13(9)17/h1-4,6-7,16-17H,5H2

InChI key

MDNWOSOZYLHTCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Antoine Ghauch et al.
Journal of hazardous materials, 164(2-3), 665-674 (2008-09-30)
Palladium, ruthenium and silver were investigated as catalysts for the dechlorination of dichlorophen (DCP, 2,2'-methylenebis(4-chlorophenol)), an antimicrobial and anthelmintic agent largely used as algicide, fungicide and bactericide. Experiments were undertaken under oxic and anoxic conditions for experimental durations up to...
J Langrand et al.
Clinical toxicology (Philadelphia, Pa.), 51(3), 178-181 (2013-03-12)
Human dichlorophen poisoning is rare. We aim to report a case of dichlorophen poisoning resulting in complete recovery despite life-threatening multiorgan failure and huge serum dichlorophen concentrations. Description of features and management in one dichlorophen-poisoned patient. After liquid-liquid extraction, dichlorophen...
P Kintz et al.
International journal of legal medicine, 110(2), 95-96 (1997-01-01)
A case is presented involving an acute fatality resulting from self-administered dichlorophen, a chlorophenol fungicide. The compound was quantified using gas chromatography/mass spectrometry after extraction with methyl-tert-butyl ether, derivatization by methylation and separation on a HP5-MS capillary column. The blood...
Corine J Houtman et al.
Environmental toxicology and chemistry, 26(5), 898-907 (2007-05-25)
The exposure to and effects of estrogenic compounds in male breams from Dutch freshwater locations were investigated. Ovotestis was observed infrequently (maximum frequency 16%). However, plasma vitellogenin (VTG) concentration was elevated highly at some locations. Estrogenic activities in male bream...
H Stouten et al.
Journal of applied toxicology : JAT, 18(4), 271-279 (1998-08-27)
As part of a health-hazard survey on the health risk of hospital cleaning workers from exposure to Lyorthol, a hazard assessment of o-benzo-p-chlorophenol, one of the constituents of Lyorthol, has been prepared. In this paper, the physical and chemical characteristics...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service