All Photos(1)

374369

Sigma-Aldrich

Hexadecafluoroheptane

technical grade, 85%

Synonym(s):
Perfluoroheptane
Linear Formula:
CF3(CF2)5CF3
CAS Number:
Molecular Weight:
388.05
Beilstein:
1716335
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

assay

85%

refractive index

n20/D 1.3 (lit.)

bp

82-84 °C (lit.)

density

1.745 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C7F16/c8-1(9,2(10,11)4(14,15)6(18,19)20)3(12,13)5(16,17)7(21,22)23

InChI key

LGUZHRODIJCVOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecafluoroheptane is a common liquid perfluoro-n-alkane. It is reported as fluorinated solvent. Encapsulation of hexadecafluoroheptane (Perfluoroheptane) by a microencapsulation method based on a Pickering emulsion templated interfacial atom transfer radical polymerization (PETI-ATRP) has been reported. Aggregation of diblock semifluorinated in perfluoroheptane (fluorinated solvent) has been reported. Physical properties and commercial/industrial uses of hexadecafluoroheptane has been reported.

Application

Hexadecafluoroheptane may be employed in solution-phase studies of perfluoropolyalkylene oligomers and in the preparation of fluorinated gel emulsions.

Packaging

10 mL in glass bottle

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Certificate of Origin

Lorenzo Tattini et al.
Journal of colloid and interface science, 339(1), 259-265 (2009-08-18)
Diblock semifluorinated n-alkanes can form aggregates and gels in fluorinated solvents. We have investigated the thermal behavior of binary mixtures comprising F(CF(2))(8)(CH(2))(16)H and fluorinated solvents. The solvents were perfluorohexane, perfluoroheptane, perfluorooctane, perfluorooctyl bromide, perfluorodecalin, and perfluorotributylamine. The phase diagrams were
Tim Brandenburg et al.
Scientific reports, 6, 31382-31382 (2016-08-17)
Research of blood substitute formulations and their base materials is of high scientific interest. Especially fluorinated microemulsions based on perfluorocarbons, with their interesting chemical properties, offer opportunities for applications in biomedicine and physical chemistry. In this work, carbon K-edge absorption
J. Chem. Phys. Phys.-Chim Biol., 91, 259-259 (1994)
Eur. Polymer J., 31, 1227-1227 (1995)
Jian Li et al.
Langmuir : the ACS journal of surfaces and colloids, 26(23), 17926-17935 (2010-10-30)
This Article describes a new microencapsulation method based on a Pickering emulsion templated interfacial atom transfer radical polymerization (PETI-ATRP). Cationic LUDOX CL nanoparticles were coated electrostatically with an anionic polymeric ATRP initiator, poly(sodium styrene sulfonate-co-2-(2-bromoisobutyryloxy)ethyl methacrylate) (PSB), prepared by radical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service