376027

Sigma-Aldrich

Ethyl 2,2,5,5-tetramethyl-1,2,5-azadisilolidin-1-acetate

97%

Synonym(s):
(1-Ethoxycarbonylmethyl)-2,2,5,5-tetramethyl-1-aza-2,5-disilacyclopentane, 2,2,5,5-Tetramethyl-1,2,5-azadisilolidin-1-acetic acid ethyl ester, STABASE-ethyl glycinate
Empirical Formula (Hill Notation):
C10H23NO2Si2
CAS Number:
Molecular Weight:
245.47
Beilstein/REAXYS Number:
3545518
MDL number:

assay

97%

refractive index

n20/D 1.451 (lit.)

bp

80-82 °C/0.25 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CN1[Si](C)(C)CC[Si]1(C)C

InChI

1S/C10H23NO2Si2/c1-6-13-10(12)9-11-14(2,3)7-8-15(11,4)5/h6-9H2,1-5H3

InChI key

NTAZFMGCNVXERO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service