MilliporeSigma
All Photos(1)

385905

Sigma-Aldrich

4-(2,3-Dihydroxypropyl) 2-isononenylsuccinate potassium salt solution

40 wt. % in H2O

Linear Formula:
HOCH2CH(OH)CH2O2CCH2CH(C9H17)CO2K
CAS Number:
Molecular Weight:
354.48
MDL number:
PubChem Substance ID:

concentration

40 wt. % in H2O

refractive index

n20/D 1.406

density

1.094 g/mL at 25 °C

SMILES string

CCCCCC\C=C(\C)C(CC(=O)OCC(O)CO)C(=O)O[K]

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service