Documents

386758

Poly-L-arginine sulfate salt

average Mv 15,000-50,000

CAS Number:
MDL number:

mol wt

average Mv 15,000-50,000

SMILES string

OS(O)(=O)=O.NC(CCCNC(N)=N)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service