387436

Sigma-Aldrich

Phenyl 1-hydroxy-2-naphthoate

99%

Synonym(s):
Phenyl 1-hydroxy-2-naphthalate, 1-Hydroxy-2-naphthoic acid phenyl ester, 2-Phenoxycarbonyl-1-naphthol, Phenyl 1-hydroxy-2-naphthoate
Linear Formula:
HOC10H6CO2C6H5
CAS Number:
Molecular Weight:
264.28
Beilstein/REAXYS Number:
2217072
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

assay

99%

form

liquid crystal

mp

94-96 °C (lit.)

SMILES string

Oc1c(ccc2ccccc12)C(=O)Oc3ccccc3

InChI

1S/C17H12O3/c18-16-14-9-5-4-6-12(14)10-11-15(16)17(19)20-13-7-2-1-3-8-13/h1-11,18H

InChI key

QHDYIMWKSCJTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.