387487

1,3-Butanediol cyclic sulfate

99%

Synonym(s):
4-Methyl-1,3,2-dioxathiane 2,2-dioxide
Empirical Formula (Hill Notation):
C4H8O4S
CAS Number:
Molecular Weight:
152.17
MDL number:

assay

99%

SMILES string

CC1CCOS(=O)(=O)O1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service