Skip to Content
MilliporeSigma

Skip To

388610

3-Heptyne

97%

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CH3CH2C≡C(CH2)2CH3
CAS Number:
Molecular Weight:
96.17
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
219-967-7
MDL number:
Assay:
97%


assay

97%

refractive index

n20/D 1.4225 (lit.)

bp

105-106 °C (lit.)

density

0.741 g/mL at 25 °C (lit.)

SMILES string

CCCC#CCC

InChI

1S/C7H12/c1-3-5-7-6-4-2/h3-5H2,1-2H3

InChI key

KLYHSJRCIZOUHE-UHFFFAOYSA-N



pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

3.2 °F - closed cup

flash_point_c

-16 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters