Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

389277

N,N-Dimethylundecylamine

97%

Synonym(s):

1-(Dimethylamino)undecane

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
CH3(CH2)10N(CH3)2
CAS Number:
Molecular Weight:
199.38
UNSPSC Code:
12352100
Beilstein/REAXYS Number:
2345290
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

MMWFTWUMBYZIRZ-UHFFFAOYSA-N

SMILES string

CCCCCCCCCCCN(C)C

InChI

1S/C13H29N/c1-4-5-6-7-8-9-10-11-12-13-14(2)3/h4-13H2,1-3H3

assay

97%

refractive index

n20/D 1.435 (lit.)

bp

50 °C/0.1 mmHg (lit.)

density

0.783 g/mL at 25 °C (lit.)

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service