391786

Sigma-Aldrich

Lithium oxide

−6-+100 mesh

Synonym(s):
Dilithium oxide, Lithium oxide, Lithia
Linear Formula:
Li2O
CAS Number:
Molecular Weight:
29.88
EC Number:
MDL number:
PubChem Substance ID:

particle size

−6-+100 mesh

density

2.013 g/mL at 25 °C (lit.)

SMILES string

[Li+].[Li+].[O--]

InChI

1S/2Li.O/q2*+1;-2

InChI key

FUJCRWPEOMXPAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

hazcat

Skin Corr. 1B

storage_class_code

8B - Non-combustible, corrosive hazardous materials

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service