393932

Sigma-Aldrich

Manganese(III) phthalocyanine chloride

Dye content 85 %

Empirical Formula (Hill Notation):
C32H16ClMnN8
CAS Number:
Molecular Weight:
602.91
MDL number:
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

composition

Dye content, 85%

λmax

726 nm

SMILES string

Cl[Mn]1N2C3N=C4N=C(N=C5N1C(=NC6N=C(N=C2C7C=CC=CC37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H18N8.ClH.Mn/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;;/h1-16,25,30H;1H;/q-2;;+3/p-1

InChI key

DQZJHBHXZYWQLQ-UHFFFAOYSA-M

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.