394289

Sigma-Aldrich

1,3,5,7-Tetramethylpyrazabole

98%

Empirical Formula (Hill Notation):
C10H18B2N4
CAS Number:
Molecular Weight:
215.90
MDL number:

assay

98%

SMILES string

C[B-]1(C)n2ccc[n+]2[B-](C)(C)[n+]3cccn13

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service