MilliporeSigma
All Photos(1)

399094

Sigma-Aldrich

Molybdenum(VI) fluoride

≥99.9%

Synonym(s):
Hexafluoromolybdenum, Molybdenum hexafluoride
Linear Formula:
MoF6
CAS Number:
Molecular Weight:
209.93
EC Number:
MDL number:
PubChem Substance ID:

assay

≥99.9%

bp

37 °C (lit.)

mp

17.5 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

SMILES string

F[Mo](F)(F)(F)(F)F

InChI

1S/6FH.Mo/h6*1H;/q;;;;;;+6/p-6

InChI key

RLCOZMCCEKDUPY-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service