Documents

399736

Sigma-Aldrich

(S)-(+)-N-[1-(1-Naphthyl)ethyl]phthalamic acid

98%

Linear Formula:
2-[C10H7CH(CH3)NHCO]C6H4CO2H
CAS Number:
Molecular Weight:
319.35
MDL number:
PubChem Substance ID:

assay

98%

optical activity

[α]25/D +48°, c = 1 in methanol

mp

167 °C (dec.) (lit.)

SMILES string

C[C@H](NC(=O)c1ccccc1C(O)=O)c2cccc3ccccc23

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service