401102

Sigma-Aldrich

Cadmium arsenide

pieces, <10 mm, 99.99%

Linear Formula:
Cd3As2
CAS Number:
Molecular Weight:
487.08
EC Number:
MDL number:

assay

99.99%

form

pieces

particle size

<10 mm

SMILES string

[Cd]=[As][Cd][As]=[Cd]

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service