Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C32H16N8Na2
CAS Number:
Molecular Weight:
558.50
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
composition
Dye content, 90%
mp
>300 °C (lit.)
λmax
649 nm, 721 nm (2nd)
SMILES string
[Na]n1c2nc3nc(nc4n([Na])c(nc5nc(nc1c6ccccc26)c7ccccc57)c8ccccc48)c9ccccc39
InChI
1S/C32H16N8.2Na/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;;/h1-16H;;/q-2;2*+1
InChI key
RLDWIFDMZUBMGW-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Leon Hamui et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-23)
In the current work, flexible diodes with flat heterojunction and dispersed heterojunction architecture were manufactured with to study the behavior of thin films of disodium phthalocyanine (Na2Pc). The thin film devices, using the electronic acceptor tetracyano-π-quinodimethane (TCNQ), were fabricated by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service