MilliporeSigma
All Photos(1)

403288

Sigma-Aldrich

Zirconium

powder, 3 μm

Synonym(s):
Zirconium element
Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
EC Number:
MDL number:
PubChem Substance ID:

form

powder

contains

solution 1:10 pentanol to water

resistivity

40 μΩ-cm, 20°C

particle size

3 μm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for a thermite reaction synthesis of a new compound, ZrXFe1-XO2, which is the first reported example of a compound containing a direct Fe-Zr bond.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service