Documents

403377

Sigma-Aldrich

Platinum-palladium-gold alloy

submicron powder

MDL number:
PubChem Substance ID:

form

submicron powder

composition

Au, 40%
Pd, 20%
Pt, 40%

SMILES string

[Pd].[Pt].[Au]

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service