410675

Sigma-Aldrich

trans-2,4-Dimethoxy-β-nitrostyrene

98%

Synonym(s):
2-(2,4-Dimethoxyphenyl)nitroethene, 1,3-Dimethoxy-4-(2-nitrovinyl)benzene
Linear Formula:
(CH3O)2C6H3CH=CHNO2
CAS Number:
Molecular Weight:
209.20
MDL number:
PubChem Substance ID:

assay

98%

mp

107-111 °C (lit.)

SMILES string

[H]\C(=C(\[H])[N+]([O-])=O)c1ccc(OC)cc1OC

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service