412538

Sigma-Aldrich

2-Chlorobenzimidazole

technical grade, 90%

Empirical Formula (Hill Notation):
C7H5ClN2
CAS Number:
Molecular Weight:
152.58
EC Number:
MDL number:
PubChem Substance ID:

grade

technical grade

assay

90%

mp

207-211 °C (lit.)

SMILES string

Clc1nc2ccccc2[nH]1

InChI

1S/C7H5ClN2/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H,9,10)

InChI key

AYPSHJCKSDNETA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service