Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Skip To
assay
97%
refractive index
n20/D 1.488 (lit.)
bp
71-73 °C/10 mmHg (lit.)
density
0.997 g/mL at 25 °C (lit.)
SMILES string
Cc1ccc(CN)o1
InChI
1S/C6H9NO/c1-5-2-3-6(4-7)8-5/h2-3H,4,7H2,1H3
InChI key
YSEAGSCGERFGBL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
134.6 °F - closed cup
flash_point_c
57 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thomas Heida et al.
Polymers, 12(8) (2020-08-13)
The demand for tailored, micrometer-scaled biomaterials in cell biology and (cell-free) biotechnology has led to the development of tunable microgel systems based on natural polymers, such as hyaluronic acid (HA). To precisely tailor their physicochemical and mechanical properties and thus
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
