Documents

416460

Sigma-Aldrich

Diisopropylamine hydrochloride

99%

Linear Formula:
[(CH3)2CH]2NH·HCl
CAS Number:
Molecular Weight:
137.65
MDL number:
PubChem Substance ID:

assay

99%

mp

216-218 °C (lit.)

SMILES string

Cl.CC(C)NC(C)C

Certificate of Analysis

Certificate of Origin

Bouclé Sébastien et al.
Molecules (Basel, Switzerland), 15(11), 7742-7749 (2010-11-03)
Treatment of 1,4,4-trimethyl-3,4-dihydroquinolin-2(1H)-one (2) with lithium diisopropylamide (LDA) followed by a wide range of electrophiles give the corresponding 4,4-dimethyl-3-substituted-3,4-dihydroquinolin-2-ones (3-13), providing a very mild electrophilic substitution of the 4,4-dimethyl-1,2,3,4-tetrahydroquinoline core.
Liyan Qiu et al.
Molecular pharmaceutics, 9(5), 1109-1117 (2012-04-13)
pH-responsive drug carriers derived from polymers containing weak base groups have been shown to improve the antitumor effect of chemotherapeutics. The common interpretation is that a "proton sponge effect" caused by pH-responsive polymers facilitates endosomal membrane destruction and accelerates cytoplasmic...
Alexander C Hoepker et al.
The Journal of organic chemistry, 76(19), 7985-7993 (2011-09-06)
Density functional theory computations [MP2/6-31G(d)//B3LYP/6-31G(d)] on the deaggregation of lithium diisopropylamide (LDA) dimer solvated by two tetrahydrofuran ligands to give the corresponding trisolvated monomer show eight structurally distinct minima. The barriers to exchange are comparable to those expected from experimental...
Decoupling deprotonation from metalation: Thia-Fries rearrangement.
Alan M Dyke et al.
Angewandte Chemie (International ed. in English), 47(27), 5067-5070 (2008-05-28)
Lekha Gupta et al.
The Journal of organic chemistry, 74(5), 2231-2233 (2009-02-05)
Ortholithiations of a range of arenes mediated by lithium diisopropylamide (LDA) in THF at -78 degrees C reveal substantial accelerations by as little as 0.5 mol % of LiCl (relative to LDA). Substrate dependencies suggest a specific range of reactivity...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service