Skip to Content
MilliporeSigma

417173

Sigma-Aldrich

Diisobutylaluminum fluoride solution

1.0 M in hexanes

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
[(CH3)2CHCH2]2AlF
CAS Number:
Molecular Weight:
160.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

concentration

1.0 M in hexanes

bp

68-70 °C

density

0.709 g/mL at 25 °C

SMILES string

CC(C)C[Al](F)CC(C)C

InChI

1S/2C4H9.Al.FH/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;1H/q;;+1;/p-1

InChI key

SJBIACKVFNCVOM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - Repr. 2 - STOT RE 2 Inhalation

target_organs

Nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service