Documents

421839

Sigma-Aldrich

(R)-(−)-3,5-Dinitro-N-(1-phenylethyl)benzamide

98%, ChiraSelect

Synonym(s):
R-(−)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
Linear Formula:
C6H5CH(CH3)NHCOC6H3(NO2)2
CAS Number:
Molecular Weight:
315.28
Beilstein/REAXYS Number:
4268701
MDL number:

product line

ChiraSelect

assay

98%

optical activity

[α]20/D −47°, c = 0.9 in acetone

optical purity

ee: ≥99% (HPLC)

mp

158-161 °C (lit.)

SMILES string

C[C@@H](NC(=O)c1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O)c2ccccc2

InChI

1S/C15H13N3O5/c1-10(11-5-3-2-4-6-11)16-15(19)12-7-13(17(20)21)9-14(8-12)18(22)23/h2-10H,1H3,(H,16,19)/t10-/m1/s1

InChI key

ABEVDCGKLRIYRW-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R) and (S) forms are reagents for the determination of the enantiomeric excesses of sulfoxides and phosphine oxides.

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Certificate of Analysis

Certificate of Origin

Tetrahedron Letters, 25, 3467-3467 (1984)
Tetrahedron Letters, 26, 2649-2649 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service