Skip to Content
MilliporeSigma

423882

Tributyltin benzoate

98%

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
C6H5CO2Sn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
411.17
UNSPSC Code:
12352103
PubChem Substance ID:
EC Number:
224-399-8
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Tributyltin benzoate, 98%

InChI key

JUEAPPHORMOWPK-UHFFFAOYSA-M

InChI

1S/C7H6O2.3C4H9.Sn/c8-7(9)6-4-2-1-3-5-6;3*1-3-4-2;/h1-5H,(H,8,9);3*1,3-4H2,2H3;/q;;;;+1/p-1

SMILES string

CCCC[Sn](CCCC)(CCCC)OC(=O)c1ccccc1

assay

98%

refractive index

n20/D 1.5182 (lit.)

density

1.193 g/mL at 25 °C (lit.)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service