MilliporeSigma
All Photos(1)

424595

Sigma-Aldrich

5-Nitro-1-pentene

98%

Linear Formula:
O2N(CH2)3CH=CH2
CAS Number:
Molecular Weight:
115.13
MDL number:
PubChem Substance ID:

assay

98%

form

liquid

refractive index

n20/D 1.44 (lit.)

bp

82 °C/32 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)CCCC=C

InChI

1S/C5H9NO2/c1-2-3-4-5-6(7)8/h2H,1,3-5H2

InChI key

NPFOLTHIKZSVPQ-UHFFFAOYSA-N

Related Categories

General description

5-Nitro-1-pentene is a nitro alkene. It undergoes conjugate addition to 3-buten-2-one by C-C bond formation. Its enthalpy of vaporization at boiling point has been determined.

Application

5-Nitro-1-pentene was used as a substrate in the microwave assisted cross metathesis reaction with allyl trimethylsilane (allylTMS).
It may be used as a precursor for the preparation of allylrethrone and as a starting material in the synthesis of 3-(3′-butenyl)-5-acetyl-2-isoxazoline.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service