425052

Sigma-Aldrich

Strontium hydroxide

99.998%

Synonym(s):
Strontium dihydroxide
Linear Formula:
Sr(OH)2
CAS Number:
Molecular Weight:
121.63
EC Number:
MDL number:

assay

99.998%

SMILES string

O[Sr]O

InChI

1S/2H2O.Sr/h2*1H2;/q;;+2/p-2

InChI key

UUCCCPNEFXQJEL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service