Skip to Content
MilliporeSigma

430617

Poly(ethylene-co-vinyl acetate-co-carbon monoxide)

average Mw >250,000 by LS, pellets

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

C=C.[C-]#[O+].COC(=O)C=C

form

pellets

melt index

35 g/10 min (190°C/2.16kg)

mol wt

average Mw >250,000 by LS

composition

carbon monoxide, 10 wt. % , vinyl acetate, 24 wt. %

hardness

70 (Shore A, 10 sec, ASTM D 2240-81)

transition temp

Tg −32 °C (ASTM D 3418-75)

density

1 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Permanent plasticizer for PVC used in upholstery, medical bags and tubing, shoes and industrial films. Adhesion promoter.

Features and Benefits

Non-migrating plasticizer, low temperature impact strength, resistant to environmental degradation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service