Skip to Content
MilliporeSigma

435619

Rhenium(VI) fluoride

99%

Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
ReF6
CAS Number:
Molecular Weight:
300.20
EC Number:
MDL number:
UNSPSC Code:
12352302

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

vapor pressure

545 mmHg ( 25 °C)

assay

99%

bp

47.6 °C (lit.)

mp

18.8 °C (lit.)

density

6 g/mL at 25 °C (lit.)

SMILES string

F[Re](F)(F)(F)(F)F

Packaging

Packaged in cylinders

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1A

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service