436410

Sigma-Aldrich

Thulium(III) chloride hydrate

99.9%

Linear Formula:
TmCl3 · xH2O
CAS Number:
Molecular Weight:
275.29 (anhydrous basis)
MDL number:

assay

99.9%

composition

Degree of hydration, ~6

SMILES string

O.Cl[Tm](Cl)Cl

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service