MilliporeSigma
All Photos(1)

437859

Sigma-Aldrich

Mercury(II) bromide

99.998%

Synonym(s):
Mercuric bromide
Linear Formula:
HgBr2
CAS Number:
Molecular Weight:
360.40
EC Number:
MDL number:
PubChem Substance ID:

vapor pressure

1 mmHg ( 136.5 °C)

assay

99.998%

form

powder

reaction suitability

reagent type: catalyst
core: mercury

bp

322 °C (lit.)

mp

236 °C (lit.)

SMILES string

Br[Hg]Br

InChI

1S/2BrH.Hg/h2*1H;/q;;+2/p-2

InChI key

NGYIMTKLQULBOO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service